Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Lcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130421.2 | NP_569105.1 | 7 | 581 | ||
GenBank mRNA | AB072980.1 | - | 7 | 421 | ||
BC100066.1 | - | 7 | 581 | 7387128 | ||
FQ232596.1 | - | 7 | 595 |