Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pou2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100639.1 | NP_001094109.1 | 5, 6 | 400 | ||
XM_008769591.1 | XP_008767813.1 | 5, 6 | 2526 | |||
XM_008769592.1 | XP_008767814.1 | 5, 6 | 661 | |||
XM_008769593.1 | XP_008767815.1 | 7, 8 | 860 | |||
XM_008769594.1 | XP_008767816.1 | 6, 7 | 762 | |||
XM_008769595.1 | XP_008767817.1 | 7, 8 | 795 | |||
XM_008769596.1 | XP_008767818.1 | 7, 8 | 2970 | |||
XM_008769597.1 | XP_008767819.1 | 6, 7 | 2872 | |||
XM_008769598.1 | XP_008767820.1 | 5, 6 | 2771 | |||
XM_008769599.1 | XP_008767821.1 | 5, 6 | 424 | |||
GenBank mRNA | DV214198.1 | - | 5, 6 | 400 |