Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other G6PD SIRNA Assays ›
Gene Symbol:
G6pd
Gene Name:
glucose-6-phosphate dehydrogenase
Gene Aliases:
G6pdx
Chromosome Location:
Chr.X: 156274800 - 156293935 on Build Rnor_6.0
UniGene ID:
Rn.11040
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS302151
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: G6pd
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017006.2
NP_058702.1
2
203
GenBank mRNA
BC081820.1
-
2
116
7108556
CB729647.1
-
2
433
X07467.1
-
2
129
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_019468, NM_008062, AK165207, Z11911, AK088135, and others.
Panther Classification:
Molecular Function -
dehydrogenase
Gene Ontology Categories:
Function(s)
Process(es)
cytokine production
angiotensin mediated vasoconstriction involved in regulation of systemic arterial blood pressure
vasodilation by angiotensin involved in regulation of systemic arterial blood pressure
glucose metabolic process
pentose-phosphate shunt
cholesterol biosynthetic process
NADP metabolic process
NADPH regeneration
NADP biosynthetic process
glutathione metabolic process
response to oxidative stress
pentose-phosphate shunt, oxidative branch
negative regulation of protein glutathionylation
response to organic cyclic compound
pentose biosynthetic process
substantia nigra development
response to food
interleukin-10 production
interleukin-12 production
cellular response to oxidative stress
regulation of multicellular organism growth
erythrocyte maturation
regulation of neuron apoptotic process
response to ethanol
ribose phosphate biosynthetic process
erythrocyte development
glucose 6-phosphate metabolic process
oxidation-reduction process
glucose-6-phosphate dehydrogenase activity
glucose binding
carbohydrate binding
identical protein binding
protein homodimerization activity
NADP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2lcrw:80/100.66.78.62:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0