Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nefm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017029.2 | NP_058725.1 | 2 | 1218 | ||
GenBank mRNA | M18628.1 | - | 2 | 1196 | ||
Z12152.1 | - | 2 | 1221 |