Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Prl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012629.1 | NP_036761.1 | 5 | 805 | ||
XM_006253912.2 | XP_006253974.1 | 5 | 697 | |||
GenBank mRNA | AF022935.1 | - | 5 | 805 | ||
BC166892.1 | - | 5 | 670 | 6919622 | ||
BC168729.1 | - | 5 | 676 | 6922061 | ||
S46794.1 | - | 4 | 541 | |||
V01249.1 | - | 5 | 670 |