Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Itga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030994.2 | NP_112256.1 | 2, 3 | 572 | ||
XM_008760735.1 | XP_008758957.1 | 2, 3 | 188 | |||
GenBank mRNA | X52140.1 | - | 2, 3 | 596 |