Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBP4 SIRNA Assays ›
Gene Symbol:
Rbp4
Gene Name:
retinol binding protein 4, plasma
Gene Aliases:
RBPA
Chromosome Location:
Chr.1: 256806476 - 256813678 on Build Rnor_6.0
UniGene ID:
Rn.108214
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS304077
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rbp4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_013162.1
NP_037294.1
2
131
GenBank mRNA
BC167099.1
-
2
132
7369314
CO561550.1
-
2
131
FQ218638.1
-
2
147
Back To Top
More Information
Panther Classification:
Molecular Function -
transfer/carrier protein
Gene Ontology Categories:
Function(s)
Process(es)
eye development
gluconeogenesis
transport
spermatogenesis
heart development
male gonad development
maintenance of gastrointestinal epithelium
lung development
positive regulation of insulin secretion
response to retinoic acid
response to insulin
retinol transport
retinol metabolic process
retinal metabolic process
glucose homeostasis
response to ethanol
embryonic organ morphogenesis
embryonic skeletal system development
cardiac muscle tissue development
female genitalia morphogenesis
detection of light stimulus involved in visual perception
positive regulation of immunoglobulin secretion
vitamin transport
retina development in camera-type eye
negative regulation of cardiac muscle cell proliferation
embryonic retina morphogenesis in camera-type eye
uterus development
vagina development
urinary bladder development
heart trabecula formation
transporter activity
retinal binding
retinol binding
retinol transporter activity
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0