Search
Search

Pde4c| Gene Symbol: Pde4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276765.1 | NP_001263694.1 | 14 | 1558 | ||
| XM_006252868.2 | XP_006252930.1 | 15 | 1800 | |||
| XM_006252869.2 | XP_006252931.1 | 13 | 1605 | |||
| GenBank mRNA | L27061.1 | - | 12 | 1154 | ||
| M25347.1 | - | 6 | 697 | |||