Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DRD3 SIRNA Assays ›
          Gene Symbol:
          Drd3
          Gene Name:
          dopamine receptor D3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 61822077 - 61883223 on Build Rnor_6.0
          UniGene ID:
          Rn.10356
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS307693
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Drd3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017140.1 NP_058836.1 2 371
          XM_006248313.2 XP_006248375.2 3 588
          GenBank mRNA X53944.1 - 2 371

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_000796, NM_033663, L20469, BC128123, BC095510, and others.
          This siRNA also targets the following transcripts in Mouse: NM_007877, BC119286, BC137819, X67274.

          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          synaptic transmission, dopaminergic
          response to amphetamine
          regulation of blood volume by renin-angiotensin
          G-protein coupled receptor internalization
          cellular calcium ion homeostasis
          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating dopamine receptor signaling pathway
          negative regulation of adenylate cyclase activity
          adenylate cyclase-inhibiting dopamine receptor signaling pathway
          dopamine receptor signaling pathway
          chemical synaptic transmission
          locomotory behavior
          positive regulation of cell proliferation
          visual learning
          regulation of dopamine secretion
          regulation of lipid metabolic process
          regulation of cAMP metabolic process
          negative regulation of sodium:proton antiporter activity
          positive regulation of cytokinesis
          circadian regulation of gene expression
          response to histamine
          gastric emptying
          positive regulation of renal sodium excretion
          regulation of locomotion
          regulation of multicellular organism growth
          response to cocaine
          response to drug
          response to morphine
          regulation of circadian sleep/wake cycle, sleep
          response to ethanol
          negative regulation of blood pressure
          positive regulation of mitotic nuclear division
          positive regulation of transcription from RNA polymerase II promoter
          acid secretion
          behavioral response to cocaine
          negative regulation of oligodendrocyte differentiation
          arachidonic acid secretion
          negative regulation of protein secretion
          musculoskeletal movement, spinal reflex action
          negative regulation of protein kinase B signaling
          prepulse inhibition
          negative regulation of dopamine receptor signaling pathway
          positive regulation of dopamine receptor signaling pathway
          regulation of locomotion involved in locomotory behavior
          dopamine neurotransmitter receptor activity, coupled via Gi/Go
          dopamine neurotransmitter receptor activity
          protein binding
          drug binding
          protein domain specific binding
          D1 dopamine receptor binding
          dopamine binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0