Search Thermo Fisher Scientific
Gene Symbol: LOC102551660 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231007.2 | XP_006231069.1 | 21, 22 | 2765 | ||
XM_006231008.1 | XP_006231070.1 | 21, 22 | 2735 | |||
XM_006231009.2 | XP_006231071.1 | 20, 21 | 2610 | |||
GenBank mRNA | FQ228620.1 | - | 21, 22 | 2735 |