Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ffar4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047088.1 | NP_001040553.1 | 2 | 649 | ||
GenBank mRNA | AB207868.1 | - | 2 | 649 |
Set Membership: |
G-protein coupled receptor |