Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017190.4 | NP_058886.1 | 7 | 1074 | ||
XM_006228824.2 | XP_006228886.1 | 6 | 1104 | |||
XM_006228825.2 | XP_006228887.1 | 6 | 1621 | |||
XM_006228827.2 | XP_006228889.1 | 7 | 1156 | |||
XM_008759137.1 | XP_008757359.1 | 6 | 1628 | |||
XM_008759138.1 | XP_008757360.1 | 6 | 1097 | |||
XM_008759139.1 | XP_008757361.1 | 6 | 1627 | |||
GenBank mRNA | FQ212027.1 | - | 7 | 1090 | ||
M14871.1 | - | 6 | 1044 | |||
M16800.1 | - | 7 | 1074 | |||
M22357.1 | - | 6 | 1022 | |||
X05301.1 | - | 6 | 1037 |