Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TGFBR1 SIRNA Assays ›
          Gene Symbol:
          Tgfbr1 LOC103690035
          Gene Name:
          transforming growth factor, beta receptor 1
          TGF-beta receptor type-1-like
          Gene Aliases:
          Alk5, Skr4, Tgfr-1, tbetaR-I
          -
          Chromosome Location:
          Chr.5: 63056079 - 63124107 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 64565362 - 64585219 on Build Rnor_6.0
          UniGene ID:
          Rn.44402
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID RSS310278
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tgfbr1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012775.2 NP_036907.2 1215
          XM_006238030.2 XP_006238092.2 1229
          GenBank mRNA BC087035.1 - 1157 7098473
          L26110.1 - 1184
          Gene Symbol: LOC103690035
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006238153.2 XP_006238215.2 6 2500
          XM_008763706.1 XP_008761928.1 6 2512

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_009370, AK015337, D25540, BC063260, D28526, and others.

          Panther Classification:

          Molecular Function -

          serine/threonine protein kinase receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          skeletal system development
          angiogenesis
          response to hypoxia
          in utero embryonic development
          kidney development
          blastocyst development
          epithelial to mesenchymal transition
          negative regulation of endothelial cell proliferation
          positive regulation of endothelial cell proliferation
          lens development in camera-type eye
          regulation of transcription, DNA-templated
          protein phosphorylation
          apoptotic process
          signal transduction
          transmembrane receptor protein serine/threonine kinase signaling pathway
          transforming growth factor beta receptor signaling pathway
          heart development
          embryo implantation
          aging
          positive regulation of cell proliferation
          germ cell migration
          male gonad development
          response to toxic substance
          post-embryonic development
          anterior/posterior pattern specification
          regulation of gene expression
          positive regulation of gene expression
          regulation of epithelial to mesenchymal transition
          positive regulation of pathway-restricted SMAD protein phosphorylation
          response to organic cyclic compound
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          signal transduction by protein phosphorylation
          collagen fibril organization
          positive regulation of cell growth
          lung development
          organ regeneration
          regulation of protein ubiquitination
          negative regulation of chondrocyte differentiation
          activin receptor signaling pathway
          response to prostaglandin E
          intracellular signal transduction
          regulation of growth
          endothelial cell activation
          negative regulation of apoptotic process
          regulation of protein binding
          endothelial cell migration
          response to estrogen
          negative regulation of endothelial cell differentiation
          positive regulation of transcription, DNA-templated
          protein autophosphorylation
          thymus development
          digestive tract development
          neuron fate commitment
          embryonic cranial skeleton morphogenesis
          skeletal system morphogenesis
          mesenchymal cell differentiation
          artery morphogenesis
          cell motility
          positive regulation of cellular component movement
          positive regulation of filopodium assembly
          response to electrical stimulus
          positive regulation of protein kinase B signaling
          parathyroid gland development
          palate development
          pharyngeal system development
          cardiac epithelial to mesenchymal transition
          pathway-restricted SMAD protein phosphorylation
          positive regulation of SMAD protein import into nucleus
          response to cholesterol
          cellular response to transforming growth factor beta stimulus
          positive regulation of apoptotic signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway
          protein kinase activity
          protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          transforming growth factor beta-activated receptor activity
          transforming growth factor beta receptor activity, type I
          receptor signaling protein activity
          type II transforming growth factor beta receptor binding
          ATP binding
          growth factor binding
          ubiquitin protein ligase binding
          protein complex binding
          SMAD binding
          metal ion binding
          protein heterodimerization activity
          transforming growth factor beta binding
          I-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ggn6l:80/100.66.76.177:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0