Search
Search

Chpf2| Gene Symbol: Chpf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106574.1 | NP_001100044.1 | 1 | 1566 | ||
| XM_006235890.2 | XP_006235952.1 | 2 | 723 | |||
| XM_006235891.2 | XP_006235953.1 | 1 | 71 | |||
| XM_008762633.1 | XP_008760855.1 | 2 | 745 | |||