Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Baat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017300.2 | NP_058996.2 | 4 | 1324 | ||
GenBank mRNA | BC088153.1 | - | 4 | 1324 | 7368634 | |
D43964.1 | - | 4 | 1339 | |||
FQ218773.1 | - | 1368 |