Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zfyve1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106743.1 | NP_001100213.1 | 4 | 1239 | ||
XM_006240323.2 | XP_006240385.1 | 4 | 1713 | |||
XM_006240324.2 | XP_006240386.1 | 4 | 1709 | |||
XM_006240325.2 | XP_006240387.1 | 4 | 1654 |