Search
Search

Mterfd1| Gene Symbol: Mterfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199387.1 | NP_955419.1 | 5 | 870 | ||
| XM_006241509.2 | XP_006241571.1 | 5 | 896 | |||
| GenBank mRNA | BC062080.1 | - | 5 | 870 | 5599618 | |
| FQ232616.1 | - | 6 | 979 | |||