Search
Search

Supt6h| Gene Symbol: Supt6h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191820.1 | NP_001178749.1 | 21 | 3046 | ||
| XM_008768006.1 | XP_008766228.1 | 21 | 2707 | |||
| XM_008768007.1 | XP_008766229.1 | 22 | 2898 | |||
| XM_008768008.1 | XP_008766230.1 | 21 | 2898 | |||