Search
Search

Supt6h| Gene Symbol: Supt6h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191820.1 | NP_001178749.1 | 27, 28 | 4052 | ||
| XM_008768006.1 | XP_008766228.1 | 27, 28 | 3713 | |||
| XM_008768007.1 | XP_008766229.1 | 28, 29 | 3904 | |||
| XM_008768008.1 | XP_008766230.1 | 27, 28 | 3904 | |||