Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mtif2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004254.1 | NP_001004254.1 | 6 | 772 | ||
GenBank mRNA | BC079421.1 | - | 6 | 772 | 7113831 | |
FQ224859.1 | - | 6 | 840 |