Search
Search

Nxf2| Gene Symbol: Nxf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002730283.4 | XP_002730329.2 | 12, 13 | 1162 | ||
| XM_006257244.2 | XP_006257306.1 | 12, 13 | 1082 | |||
| XM_006257245.2 | XP_006257307.1 | 12, 13 | 1075 | |||