Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rnf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107929.2 | NP_001101399.2 | 15 | 2539 | ||
XM_006238154.2 | XP_006238216.1 | 16 | 2464 | |||
XM_006238155.2 | XP_006238217.1 | 16 | 2383 |