Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Npas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108214.2 | NP_001101684.2 | 13 | 1713 | ||
XM_008767031.1 | XP_008765253.1 | 12 | 1188 | |||
XM_008767032.1 | XP_008765254.1 | 13 | 2439 | |||
XM_008767033.1 | XP_008765255.1 | 13 | 1903 | |||
XM_008767034.1 | XP_008765256.1 | 13 | 2796 | |||
GenBank mRNA | BC168989.1 | - | 5 | 516 |