Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Itgb1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108245.2 | NP_001101715.1 | 6, 7 | 521 | ||
GenBank mRNA | FM116382.1 | - | 6, 7 | 480 | ||
FM118664.1 | - | 6, 7 | 505 | |||
FQ215476.1 | - | 6, 7 | 516 | |||
FQ216044.1 | - | 6, 7 | 520 | |||
FQ216100.1 | - | 6, 7 | 497 | |||
FQ216185.1 | - | 6, 7 | 517 | |||
FQ224925.1 | - | 6, 7 | 518 |