Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Habp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079940.1 | NP_001073409.1 | 2 | 463 | ||
XM_006253494.2 | XP_006253556.1 | 2 | 518 | |||
GenBank mRNA | BC129111.1 | - | 2 | 463 | 7111244 |