Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Agmo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135899.1 | NP_001129371.1 | 6, 7 | 810 | ||
GenBank mRNA | BC127545.1 | - | 6, 7 | 810 | 7321962 | |
FQ211046.1 | - | 6, 7 | 866 | |||
FQ219104.1 | - | 6, 7 | 854 |