Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zbtb44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034942.1 | NP_001030114.1 | 5 | 1446 | ||
XM_006242768.2 | XP_006242830.1 | 5 | 1633 | |||
XM_006242769.2 | XP_006242831.1 | 5 | 1579 | |||
GenBank mRNA | BC104680.1 | - | 5 | 1446 | 7456912 |