Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ick | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138886.1 | NP_620241.1 | 12 | 1853 | ||
XM_008766345.1 | XP_008764567.1 | 12 | 2076 | |||
GenBank mRNA | D26178.1 | - | 12 | 1853 |