Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mbtps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053569.1 | NP_446021.1 | 18 | 2799 | ||
XM_006255723.2 | XP_006255785.1 | 17 | 4031 | |||
XM_008772651.1 | XP_008770873.1 | 17 | 4031 | |||
GenBank mRNA | AF094821.1 | - | 18 | 2799 | ||
BC061855.1 | - | 17 | 2708 |