Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rhbdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191822.1 | NP_001178751.1 | 8 | 1031 | ||
XM_006245894.2 | XP_006245956.1 | 7 | 2611 | |||
XM_008767537.1 | XP_008765759.1 | 8 | 2785 | |||
GenBank mRNA | Y17258.1 | - | 5 | 415 |