Search
Search

Tbpl1| Gene Symbol: Tbpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006227721.2 | XP_006227783.1 | 6 | 785 | ||
| XM_006227722.2 | XP_006227784.1 | 6 | 709 | |||
| XM_008758627.1 | XP_008756849.1 | 7 | 752 | |||
| GenBank mRNA | CF109177.1 | - | 5 | 444 | ||