Search
Search

Noxo1| Gene Symbol: Noxo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106986.1 | NP_001100456.1 | 7 | 1494 | ||
| XM_006245948.1 | XP_006246010.1 | 8 | 887 | |||
| XM_006245949.1 | XP_006246011.1 | 6, 7 | 1208 | |||
| XM_006245950.2 | XP_006246012.1 | 7 | 1274 | |||
| XM_008767566.1 | XP_008765788.1 | 7 | 1274 | |||