Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135033.2 | NP_001128505.1 | 4 | 973 | ||
XM_008767824.1 | XP_008766046.1 | 4 | 1234 |