Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Thoc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271293.1 | NP_001258222.1 | 5 | 438 | ||
NM_001271294.1 | NP_001258223.1 | 5 | 482 | |||
XM_006251724.1 | XP_006251786.1 | 5 | 562 | |||
XM_006251725.1 | XP_006251787.1 | 5 | 449 | |||
GenBank mRNA | EV771598.1 | - | 5 | 484 | ||
FM110383.1 | - | 5 | 398 |