Search Thermo Fisher Scientific
Gene Symbol: Akr1e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008342.1 | NP_001008343.1 | 2 | 259 | ||
XM_008771865.1 | XP_008770087.1 | 2 | 279 | |||
GenBank mRNA | BC086397.1 | - | 2 | 259 | 7302893 | |
FQ212583.1 | - | 2 | 272 |