Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Irs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032074.1 | NP_114463.1 | 1 | 489 | ||
GenBank mRNA | BC161840.1 | - | 1 | 341 | 9027966 | |
U93880.1 | - | 1 | 489 |