Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FTL1 SIRNA Assays ›
          Gene Symbol:
          Ftl1 Fam65b LOC100360087
          Gene Name:
          ferritin light chain 1
          family with sequence similarity 65, member B
          ferritin light chain 1-like
          Gene Aliases:
          Ftl
          Ab2-162, RGD1306939
          -
          Chromosome Location:
          Chr.1: 101448190 - 101450034 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 42324594 - 42444935 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 152325856 - 152326797 on Build Rnor_6.0
          UniGene ID:
          Rn.1905
          Rn.202381
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID RSS340967
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ftl1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_022500.4 NP_071945.3 4 717
          GenBank mRNA BC061525.1 - 4 721 6921603
          BC086583.1 - 4 698 7307718
          BC088756.1 - 4 698 7307747
          FQ209407.1 - 4 717
          FQ209694.1 - 4 717
          FQ218577.1 - 4 717
          FQ219483.1 - 4 717
          FQ219809.1 - 4 717
          FQ220095.1 - 4 717
          FQ220123.1 - 4 717
          FQ220243.1 - 4 714
          FQ220651.1 - 4 717
          FQ220884.1 - 4 717
          FQ221898.1 - 4 717
          FQ222411.1 - 3 621
          FQ223227.1 - 4 717
          FQ226254.1 - 4 673
          FQ226458.1 - 4 716
          FQ226986.1 - 4 713
          FQ227401.1 - 4 717
          FQ227800.1 - 4 717
          FQ227843.1 - 4 719
          FQ228050.1 - 4 717
          FQ228585.1 - 4 717
          FQ229176.1 - 4 717
          FQ229284.1 - 4 717
          FQ229370.1 - 4 717
          FQ229673.1 - 4 715
          FQ229757.1 - 4 717
          FQ230098.1 - 4 715
          FQ230445.1 - 4 717
          FQ230659.1 - 4 713
          FQ231322.1 - 4 717
          FQ231373.1 - 4 717
          FQ231415.1 - 3 573
          FQ231464.1 - 4 717
          FQ231570.1 - 4 717
          FQ231615.1 - 4 717
          FQ231781.1 - 4 713
          FQ232031.1 - 4 717
          FQ232167.1 - 4 717
          FQ232517.1 - 4 717
          FQ233772.1 - 4 718
          FQ233996.1 - 4 717
          FQ234074.1 - 4 717
          FQ234647.1 - 4 717
          FQ235250.1 - 4 717
          K01930.1 - 4 642
          L01122.1 - 4 670
          Gene Symbol: Fam65b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001014009.2 NP_001014031.2 27 3893
          GenBank mRNA AY325194.1 - 27 3893
          Gene Symbol: LOC100360087
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_002729553.4 XP_002729599.1 1 732

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: S89400.

          Panther Classification:

          Molecular Function -

          storage protein non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          iron ion transport
          cellular iron ion homeostasis
          response to lead ion
          sensory perception of sound
          molecular_function
          iron ion binding
          ferric iron binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0