Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Thap7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105863.2 | NP_001099333.2 | 4 | 311 | ||
XM_008768864.1 | XP_008767086.1 | 3 | 294 | |||
GenBank mRNA | AW916308.1 | - | 4 | 311 | ||
BC083814.1 | - | 1 | 1949 | |||
BC168998.1 | - | 3 | 233 | 7387094 |