Search Thermo Fisher Scientific
Gene Symbol: Atf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012912.2 | NP_037044.1 | 4 | 619 | ||
GenBank mRNA | BC078903.1 | - | 4 | 619 | 7100767 | |
M63282.1 | - | 4 | 624 |