Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL4 SIRNA Assays ›
          Gene Symbol:
          Il4
          Gene Name:
          interleukin 4
          Gene Aliases:
          Il4e12
          Chromosome Location:
          Chr.10: 38963979 - 38969531 on Build Rnor_6.0
          UniGene ID:
          Rn.108255
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS352748
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Il4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_201270.1 NP_958427.1 2 185
          GenBank mRNA AY496861.1 - 395
          X16058.2 - 2 185

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          interleukin superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          microglial cell activation
          positive regulation of protein phosphorylation
          innate immune response in mucosa
          positive regulation of defense response to virus by host
          T-helper 1 cell lineage commitment
          positive regulation of immunoglobulin production
          negative regulation of acute inflammatory response
          negative regulation of chronic inflammatory response
          female pregnancy
          response to nutrient
          cholesterol metabolic process
          positive regulation of cell proliferation
          regulation of proton transport
          response to organic cyclic compound
          positive regulation of B cell proliferation
          B cell costimulation
          positive regulation of interleukin-10 production
          positive regulation of interleukin-13 production
          response to cytokine
          T-helper 2 cell cytokine production
          positive regulation of T cell proliferation
          positive regulation of activated T cell proliferation
          B cell activation
          response to drug
          positive regulation of tyrosine phosphorylation of Stat5 protein
          defense response to protozoan
          negative regulation of macrophage activation
          positive regulation of mast cell degranulation
          negative regulation of nitric oxide biosynthetic process
          T-helper 2 cell differentiation
          positive regulation of chemokine biosynthetic process
          positive regulation of MHC class II biosynthetic process
          response to ethanol
          positive regulation of T cell differentiation
          negative regulation of osteoclast differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of isotype switching to IgE isotypes
          positive regulation of isotype switching to IgG isotypes
          regulation of inflammatory response
          positive regulation of peptidyl-tyrosine phosphorylation
          regulation of immune response
          regulation of T cell activation
          negative regulation of T cell activation
          positive regulation of B cell activation
          positive regulation of sequence-specific DNA binding transcription factor activity
          retina development in camera-type eye
          cellular response to mercury ion
          positive regulation of mononuclear cell migration
          dendritic cell differentiation
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of myoblast fusion
          negative regulation of complement-dependent cytotoxicity
          negative regulation of T-helper 17 cell differentiation
          negative regulation of endothelial cell apoptotic process
          positive regulation of eosinophil chemotaxis
          negative regulation of extrinsic apoptotic signaling pathway
          cytokine activity
          interleukin-4 receptor binding
          growth factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0