Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IL4 SIRNA Assays ›
Gene Symbol:
Il4
Gene Name:
interleukin 4
Gene Aliases:
Il4e12
Chromosome Location:
Chr.10: 38963979 - 38969531 on Build Rnor_6.0
UniGene ID:
Rn.108255
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS352750
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Il4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_201270.1
NP_958427.1
1
93
GenBank mRNA
AY496861.1
-
1
63
X16058.2
-
1
93
Back To Top
More Information
Panther Classification:
Molecular Function -
interleukin superfamily
Gene Ontology Categories:
Function(s)
Process(es)
microglial cell activation
positive regulation of protein phosphorylation
innate immune response in mucosa
positive regulation of defense response to virus by host
T-helper 1 cell lineage commitment
positive regulation of immunoglobulin production
negative regulation of acute inflammatory response
negative regulation of chronic inflammatory response
female pregnancy
response to nutrient
cholesterol metabolic process
positive regulation of cell proliferation
regulation of proton transport
response to organic cyclic compound
positive regulation of B cell proliferation
B cell costimulation
positive regulation of interleukin-10 production
positive regulation of interleukin-13 production
response to cytokine
T-helper 2 cell cytokine production
positive regulation of T cell proliferation
positive regulation of activated T cell proliferation
B cell activation
response to drug
positive regulation of tyrosine phosphorylation of Stat5 protein
defense response to protozoan
negative regulation of macrophage activation
positive regulation of mast cell degranulation
negative regulation of nitric oxide biosynthetic process
T-helper 2 cell differentiation
positive regulation of chemokine biosynthetic process
positive regulation of MHC class II biosynthetic process
response to ethanol
positive regulation of T cell differentiation
negative regulation of osteoclast differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of isotype switching to IgE isotypes
positive regulation of isotype switching to IgG isotypes
regulation of inflammatory response
positive regulation of peptidyl-tyrosine phosphorylation
regulation of immune response
regulation of T cell activation
negative regulation of T cell activation
positive regulation of B cell activation
positive regulation of sequence-specific DNA binding transcription factor activity
retina development in camera-type eye
cellular response to mercury ion
positive regulation of mononuclear cell migration
dendritic cell differentiation
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of myoblast fusion
negative regulation of complement-dependent cytotoxicity
negative regulation of T-helper 17 cell differentiation
negative regulation of endothelial cell apoptotic process
positive regulation of eosinophil chemotaxis
negative regulation of extrinsic apoptotic signaling pathway
cytokine activity
interleukin-4 receptor binding
growth factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-znbv5:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0