Search
Search

Cux2| Gene Symbol: Cux2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271380.1 | NP_001258309.1 | 22 | 3871 | ||
| XM_008769215.1 | XP_008767437.1 | 17 | 3516 | |||
| XM_008769216.1 | XP_008767438.1 | 17 | 3495 | |||
| XM_008769217.1 | XP_008767439.1 | 17 | 3483 | |||
| XM_008769218.1 | XP_008767440.1 | 17 | 3462 | |||
| XM_008769219.1 | XP_008767441.1 | 21 | 4972 | |||