Search
Search

Pde4c| Gene Symbol: Pde4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276765.1 | NP_001263694.1 | 9, 10 | 977 | ||
| XM_006252868.2 | XP_006252930.1 | 10, 11 | 1219 | |||
| XM_006252869.2 | XP_006252931.1 | 8, 9 | 1024 | |||
| GenBank mRNA | L27061.1 | - | 7, 8 | 573 | ||
| M25347.1 | - | 1, 2 | 116 | |||