Search
Search

Apoc2| Gene Symbol: Apoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085352.1 | NP_001078821.1 | 3 | 192 | ||
| GenBank mRNA | BM385272.1 | - | 2 | 325 | ||
| FQ210391.1 | - | 3 | 201 | |||
| FQ210737.1 | - | 3 | 202 | |||
| FQ211227.1 | - | 3 | 202 | |||
| FQ212251.1 | - | 3 | 202 | |||
| FQ218101.1 | - | 3 | 202 | |||
| FQ218174.1 | - | 3 | 202 | |||
| FQ218270.1 | - | 3 | 201 | |||