Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pbrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252685.2 | XP_006252747.1 | 18 | 2736 | ||
XM_006252686.2 | XP_006252748.1 | 17 | 1998 | |||
XM_006252696.2 | XP_006252758.1 | 16 | 1874 | |||
XM_008771034.1 | XP_008769256.1 | 17 | 2944 | |||
XM_008771035.1 | XP_008769257.1 | 17 | 2346 | |||
XM_008771036.1 | XP_008769258.1 | 17 | 2941 | |||
XM_008771037.1 | XP_008769259.1 | 17 | 2941 | |||
XM_008771038.1 | XP_008769260.1 | 17 | 2935 | |||
XM_008771039.1 | XP_008769261.1 | 17 | 2932 | |||
XM_008771040.1 | XP_008769262.1 | 16 | 2899 | |||
XM_008771041.1 | XP_008769263.1 | 16 | 2896 | |||
XM_008771042.1 | XP_008769264.1 | 17 | 2612 | |||
XM_008771043.1 | XP_008769265.1 | 17 | 2609 | |||
XM_008771044.1 | XP_008769266.1 | 16 | 2848 | |||
XM_008771045.1 | XP_008769267.1 | 17 | 2944 | |||
XM_008771046.1 | XP_008769268.1 | 17 | 2941 | |||
XM_008771047.1 | XP_008769269.1 | 17 | 2944 | |||
XM_008771048.1 | XP_008769270.1 | 16 | 2896 | |||
XM_008771049.1 | XP_008769271.1 | 15 | 2800 | |||
XM_008771050.1 | XP_008769272.1 | 17 | 2944 | |||
XM_008771051.1 | XP_008769273.1 | 17 | 2941 | |||
XM_008771052.1 | XP_008769274.1 | 16 | 2896 |