Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap3b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748891.3 | XP_003748939.1 | 26 | 3524 | ||
XM_006229468.2 | XP_006229530.1 | 26 | 3558 | |||
XM_006229469.2 | XP_006229531.1 | 26 | 3529 |