Search
Search

Ubash3a| Gene Symbol: Ubash3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107620.1 | NP_001101090.1 | 6 | 1134 | ||
| XM_006256215.2 | XP_006256277.1 | 7 | 1258 | |||
| XM_008772818.1 | XP_008771040.1 | 7 | 874 | |||
| XM_008772819.1 | XP_008771041.1 | 7 | 869 | |||