Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly49i7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009500.1 | NP_001009500.1 | 4 | 515 | ||
XM_008763320.1 | XP_008761542.1 | 5 | 1612 | |||
GenBank mRNA | AY659934.1 | - | 4 | 515 |
Gene Symbol: LOC100911272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763440.1 | XP_008761662.1 | 5 | 1371 |