Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113542.1 | NP_001107014.1 | 3, 4 | 287 | ||
NM_001113543.1 | NP_001107015.1 | 3, 4 | 287 | |||
NM_001113544.1 | NP_001107016.1 | 3, 4 | 221 | |||
XM_006227892.2 | XP_006227954.1 | 3, 4 | 267 | |||
XM_006227893.2 | XP_006227955.1 | 3, 4 | 445 | |||
GenBank mRNA | CB613741.1 | - | 3, 4 | 216 | ||
CB613959.1 | - | 3, 4 | 221 | |||
CK478322.1 | - | 3, 4 | 287 | |||
CV108433.1 | - | 3, 4 | 284 | |||
FQ228682.1 | - | 3, 4 | 326 |