Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gabrq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031733.1 | NP_113921.1 | 5 | 534 | ||
XM_006229516.2 | XP_006229578.1 | 6 | 698 | |||
GenBank mRNA | AF419333.1 | - | 5 | 534 |